Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRT2
Entrez Gene ID
10655
Gene Name
doublesex and mab-3 related transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.9: 1003237 - 1057554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130865.2 NP_001124337.1 2 - 3 283 150
NM_006557.6 NP_006548.1 1 - 2 428 150
NM_181872.4 NP_870987.2 1 - 2 154 150
XM_011517687.1 XP_011515989.1 3 - 4 2702 150
XM_011517690.2 XP_011515992.1 2 - 3 782 150
XM_017014213.1 XP_016869702.1 3 - 4 1104 150
XM_017014214.1 XP_016869703.1 2 - 3 779 150
XM_017014216.1 XP_016869705.1 1 - 2 916 150
GenBank mRNA AF130729.1 - 1 - 2 173 150
BC030204.1 - 2 - 3 283 150
Y19052.1 - 1 - 2 408 150

View Full Product Details