Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PARVG
- Entrez Gene ID
-
64098
- Gene Name
- parvin gamma
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 44170228 - 44208469 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU584188.1 |
- |
1 - 2 |
181 |
|
Chr. 22 - 44173008 - 44183360 [+] on Build GRCh38 |
76 |
GenBankmRNA |
BU584196.1 |
- |
1 - 2 |
187 |
|
Chr. 22 - 44173002 - 44183406 [+] on Build GRCh38 |
76 |
GenBankmRNA |
DA041161.1 |
- |
1 - 2 |
190 |
|
Chr. 22 - 44172999 - 44186785 [+] on Build GRCh38 |
76 |
GenBankmRNA |
DA467313.1 |
- |
1 - 2 |
205 |
|
Chr. 22 - 44172985 - 44186783 [+] on Build GRCh38 |
76 |
GenBankmRNA |
BP299715.1 |
- |
1 - 2 |
205 |
|
Chr. 22 - 44172984 - 44189225 [+] on Build GRCh38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
parvin beta |
PARVB |
CGI-56 |
-3.8kb
|
Sense |
Chr. - 43999211 - 44169233 on Build GRCh38 |
Downstream |
KIAA1644 |
KIAA1644 |
|
60.3kb
|
Anti-sense |
Chr. - 44243665 - 44331693 on Build GRCh38 |
View Full Product Details