Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAMD4
Entrez Gene ID
23151
Gene Name
GRAM domain containing 4
Gene Aliases
DIP, dA59H18.1, dJ439F8.1
Chromosome Location
Chr.22: 46576017 - 46682755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015124.4 NP_055939.1 14 - 15 1452 61
XM_005261396.3 XP_005261453.1 14 - 15 1388 61
XM_006724169.1 XP_006724232.1 14 - 15 1350 61
XM_006724170.3 XP_006724233.1 14 - 15 1369 61
XM_011530011.1 XP_011528313.1 13 - 14 1288 61
XM_011530012.1 XP_011528314.1 13 - 14 1288 61
XM_011530013.2 XP_011528315.2 14 - 15 1397 61
XM_017028670.1 XP_016884159.1 14 - 15 1410 61
GenBank mRNA AB018310.2 - 14 - 15 1452 61
BC129837.1 - 13 - 14 1273 61
CR456470.1 - 13 - 14 1278 61

View Full Product Details