The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184782.1 | NP_001171711.1 | 4 - 5 | 1244 | 116 |
NM_018015.5 | NP_060485.4 | 4 - 5 | 1244 | 116 | |
GenBank mRNA | AK001040.1 | - | 4 - 5 | 1199 | 116 |
AK024253.1 | - | 4 - 5 | 518 | 116 | |
AK291730.1 | - | 4 - 5 | 1193 | 116 | |
BC011483.1 | - | 4 - 5 | 1165 | 116 | |
BC033709.1 | - | 4 - 5 | 1156 | 116 | |
BC054032.2 | - | 4 - 5 | 1132 | 116 | |
BC070110.1 | - | 4 - 5 | 1195 | 116 |