The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184782.1 | NP_001171711.1 | 8 - 9 | 1592 | 55 |
GenBank mRNA | AK024253.1 | - | 8 - 9 | 866 | 55 |