The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000870.6 | NP_000861.1 | 4 - 5 | 867 | 111 |
NM_001040169.2 | NP_001035259.1 | 3 - 4 | 401 | 111 | |
NM_001040172.2 | NP_001035262.2 | 3 - 4 | 401 | 111 | |
NM_001040173.2 | NP_001035263.1 | 3 - 4 | 401 | 111 | |
NM_001286410.1 | NP_001273339.1 | 3 - 4 | 401 | 111 | |
NM_199453.3 | NP_955525.1 | 3 - 4 | 401 | 111 | |
NR_104445.1 | 4 - 5 | 867 | 111 | ||
GenBank mRNA | AJ011371.1 | - | 3 - 4 | 356 | 111 |
AJ131724.1 | - | 4 - 5 | 494 | 111 | |
AJ131725.1 | - | 3 - 4 | 360 | 111 | |
AJ131726.1 | - | 3 - 4 | 360 | 111 | |
AJ278979.1 | - | 3 - 4 | 360 | 111 | |
AJ278980.1 | - | 3 - 4 | 360 | 111 | |
AJ278981.1 | - | 3 - 4 | 360 | 111 | |
AJ278982.1 | - | 3 - 4 | 360 | 111 | |
AJ519673.1 | - | 3 - 4 | 373 | 111 | |
AJ633645.1 | - | 3 - 4 | 354 | 111 | |
AK308471.1 | - | 4 - 5 | 867 | 111 | |
AM712912.1 | - | 3 - 4 | 354 | 111 | |
BC074755.2 | - | 3 - 4 | 395 | 111 | |
BC095497.1 | - | 3 - 4 | 360 | 111 | |
DR000612.1 | - | 4 - 5 | 524 | 111 | |
Y08756.1 | - | 3 - 4 | 372 | 111 | |
Y09586.1 | - | 3 - 4 | 356 | 111 | |
Y10437.1 | - | 3 - 4 | 354 | 111 | |
Y12505.1 | - | 3 - 4 | 372 | 111 | |
Y12506.1 | - | 3 - 4 | 372 | 111 | |
Y12507.1 | - | 3 - 4 | 372 | 111 | |
Y13584.1 | - | 3 - 4 | 354 | 111 |