The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184697.1 | NP_001171626.1 | 3 - 4 | 160 | 118 |
NM_001291183.1 | NP_001278112.1 | 2 - 3 | 114 | 118 | |
XM_006714278.2 | XP_006714341.1 | 3 - 4 | 197 | 118 | |
GenBank mRNA | AK307774.1 | - | 3 - 4 | 160 | 118 |
DQ854717.1 | - | 2 - 3 | 77 | 118 |