Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGEF1B
Entrez Gene ID
153020
Gene Name
RasGEF domain family member 1B
Gene Aliases
GPIG4
Chromosome Location
Chr.4: 81426393 - 81471928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300735.1 NP_001287664.1 6 - 7 886 65
NM_001300736.1 NP_001287665.1 5 - 6 763 65
NM_152545.2 NP_689758.1 6 - 7 889 65
XM_017007812.1 XP_016863301.1 4 - 5 607 65
XM_017007813.1 XP_016863302.1 6 - 7 977 65
GenBank mRNA AK056257.1 - 6 - 7 868 65
AK291459.1 - 6 - 7 834 65
AK303625.1 - 5 - 6 738 65
BC121003.1 - 6 - 7 801 65
BC121004.1 - 5 - 6 678 65

View Full Product Details