Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGEF1B
Entrez Gene ID
153020
Gene Name
RasGEF domain family member 1B
Gene Aliases
GPIG4
Chromosome Location
Chr.4: 81426393 - 81471928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300735.1 NP_001287664.1 9 - 10 1167 93
NM_001300736.1 NP_001287665.1 8 - 9 1044 93
NM_152545.2 NP_689758.1 9 - 10 1170 93
XM_017007812.1 XP_016863301.1 7 - 8 888 93
XM_017007813.1 XP_016863302.1 9 - 10 1258 93
GenBank mRNA AK056257.1 - 9 - 10 1149 93
AK291459.1 - 9 - 10 1115 93
AK303625.1 - 8 - 9 1019 93
BC121003.1 - 9 - 10 1082 93
BC121004.1 - 8 - 9 959 93

View Full Product Details