The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145933.1 | NP_001139405.1 | 9 - 10 | 1486 | 112 |
NM_001145934.1 | NP_001139406.1 | 8 - 9 | 1431 | 112 | |
NM_012253.3 | NP_036385.3 | 9 - 10 | 1504 | 112 | |
GenBank mRNA | AK292224.1 | - | 9 - 10 | 1376 | 112 |
AK292261.1 | - | 9 - 10 | 1380 | 112 | |
AK302044.1 | - | 9 - 10 | 1362 | 112 | |
AK302261.1 | - | 5 - 6 | 769 | 112 | |
BC025382.2 | - | 9 - 10 | 1380 | 112 | |
X91817.1 | - | 8 - 9 | 1270 | 112 |