The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164462.1 | NP_001157934.1 | 2 - 3 | 14952 | 80 |
GenBank mRNA | AF147790.1 | - | 1 - 2 | 704 | 80 |
AK024856.1 | - | 1 - 2 | 459 | 80 | |
BC127877.1 | - | 1 - 2 | 722 | 80 |