The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057092.2 | NP_476433.1 | 1 - 2 | 99 | 79 |
GenBank mRNA | BC003384.1 | - | 1 - 2 | 99 | 79 |