Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DERL2
Entrez Gene ID
51009
Gene Name
derlin 2
Gene Aliases
CGI-101, DERtrin-2, F-LAN-1, F-LANa, FLANa
Chromosome Location
Chr.17: 5471251 - 5486230 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304777.1 NP_001291706.1 5 - 6 587 113
NM_016041.4 NP_057125.2 5 - 6 590 113
NR_130905.1 4 - 5 702 113
NR_130906.1 4 - 5 496 113
GenBank mRNA AF132289.1 - 5 - 6 538 113
AF151859.1 - 5 - 6 528 113
AF208065.1 - 5 - 6 526 113
AF242523.1 - 5 - 6 526 113
AK024574.1 - 1 - 2 208 113
BC010890.1 - 5 - 6 538 113
CN362881.1 - 5 - 6 508 113
CN362882.1 - 4 - 5 414 113
CR457202.1 - 5 - 6 521 113

View Full Product Details