The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003259.3 | NP_003250.3 | 8 - 9 | 2060 | 70 |
XM_011528229.1 | XP_011526531.1 | 9 - 10 | 2132 | 70 | |
XM_017027185.1 | XP_016882674.1 | 5 - 6 | 1126 | 70 | |
GenBank mRNA | AX746344.1 | - | 8 - 9 | 2034 | 70 |
BC026338.1 | - | 8 - 9 | 2058 | 70 | |
BC030132.2 | - | 6 - 7 | 1659 | 70 | |
U72671.1 | - | 8 - 9 | 2060 | 70 |