Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEST1
Entrez Gene ID
7439
Gene Name
bestrophin 1
Gene Aliases
ARB, BEST, BMD, RP50, TU15B, VMD2
Chromosome Location
Chr.11: 61946722 - 61964465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004183.3 NP_004174.1 2 - 3 736 85
NR_134580.1 2 - 3 736 85
XM_005274210.3 XP_005274267.1 2 - 3 846 85
XM_005274219.3 XP_005274276.1 2 - 3 846 85
XM_005274221.3 XP_005274278.1 2 - 3 846 85
XM_011545229.2 XP_011543531.1 2 - 3 286 85
XM_017018229.1 XP_016873718.1 2 - 3 846 85
GenBank mRNA AF057169.1 - 2 - 3 260 85
AF057170.1 - 2 - 3 260 85
AF073501.1 - 2 - 3 244 85
AK289681.1 - 2 - 3 264 85
AK294577.1 - 2 - 3 264 85
AK295580.1 - 2 - 3 269 85
AY515704.1 - 1 - 2 156 85

View Full Product Details