Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRB10
Entrez Gene ID
2887
Gene Name
growth factor receptor bound protein 10
Gene Aliases
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location
Chr.7: 50590063 - 50793462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001549.2 NP_001001549.1 5 - 6 692 109
NM_001001550.2 NP_001001550.1 5 - 6 772 109
NM_001001555.2 NP_001001555.1 6 - 7 1019 109
NM_005311.4 NP_005302.3 5 - 6 692 109
XM_011515304.1 XP_011513606.1 8 - 9 1131 109
XM_011515312.2 XP_011513614.1 7 - 8 1419 109
XM_011515316.1 XP_011513618.1 6 - 7 964 109
XM_011515323.2 XP_011513625.1 3 - 4 2468 109
XM_017012029.1 XP_016867518.1 7 - 8 1351 109
XM_017012030.1 XP_016867519.1 6 - 7 1285 109
XM_017012031.1 XP_016867520.1 6 - 7 1216 109
XM_017012032.1 XP_016867521.1 12 - 13 1728 109
XM_017012033.1 XP_016867522.1 9 - 10 1266 109
XM_017012034.1 XP_016867523.1 7 - 8 867 109
XM_017012035.1 XP_016867524.1 13 - 14 2777 109
XM_017012036.1 XP_016867525.1 14 - 15 2562 109
XM_017012037.1 XP_016867526.1 14 - 15 2399 109
XM_017012038.1 XP_016867527.1 13 - 14 2349 109
XM_017012039.1 XP_016867528.1 10 - 11 1596 109
XM_017012040.1 XP_016867529.1 10 - 11 2132 109
XM_017012041.1 XP_016867530.1 12 - 13 2253 109
XM_017012042.1 XP_016867531.1 8 - 9 1173 109
XM_017012043.1 XP_016867532.1 9 - 10 1363 109
XM_017012044.1 XP_016867533.1 5 - 6 962 109
XM_017012045.1 XP_016867534.1 5 - 6 626 109
XM_017012046.1 XP_016867535.1 9 - 10 1978 109
XM_017012047.1 XP_016867536.1 14 - 15 2961 109
XM_017012048.1 XP_016867537.1 13 - 14 2845 109
XM_017012049.1 XP_016867538.1 14 - 15 2467 109
XM_017012050.1 XP_016867539.1 13 - 14 2417 109
XM_017012051.1 XP_016867540.1 12 - 13 2321 109
XM_017012052.1 XP_016867541.1 12 - 13 2661 109
XM_017012053.1 XP_016867542.1 13 - 14 2283 109
XM_017012054.1 XP_016867543.1 12 - 13 2233 109
XM_017012055.1 XP_016867544.1 9 - 10 2016 109
XM_017012056.1 XP_016867545.1 8 - 9 2848 109
XM_017012057.1 XP_016867546.1 11 - 12 2137 109
XM_017012058.1 XP_016867547.1 7 - 8 1008 109
XM_017012059.1 XP_016867548.1 7 - 8 1130 109
XM_017012060.1 XP_016867549.1 15 - 16 3094 109
XM_017012061.1 XP_016867550.1 6 - 7 960 109
XM_017012062.1 XP_016867551.1 5 - 6 721 109
XM_017012063.1 XP_016867552.1 13 - 14 2826 109
XM_017012064.1 XP_016867553.1 12 - 13 2710 109
XM_017012065.1 XP_016867554.1 10 - 11 2162 109
XM_017012066.1 XP_016867555.1 8 - 9 2521 109
XM_017012067.1 XP_016867556.1 8 - 9 1024 109
XM_017012068.1 XP_016867557.1 8 - 9 1890 109
GenBank mRNA AB000731.1 - 5 - 6 667 109
AF000017.1 - 5 - 6 661 109
AF001534.1 - 5 - 6 692 109
AJ271366.1 - 8 - 9 1131 109
AK289511.1 - 7 - 8 986 109
BC024285.1 - 6 - 7 796 109
BX649028.1 - 5 - 6 752 109
D86962.1 - 7 - 8 1424 109
U34355.1 - 5 - 6 692 109
U66065.1 - 5 - 6 772 109
U69276.1 - 5 - 6 774 109

View Full Product Details