The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000121.3 | NP_000112.1 | 7 - 8 | 1047 | 60 |
NR_033663.1 | 7 - 8 | 1297 | 60 | ||
GenBank mRNA | AK315097.1 | - | 7 - 8 | 1015 | 60 |
BC112153.1 | - | 7 - 8 | 989 | 60 | |
BC143697.1 | - | 8 - 9 | 1125 | 60 | |
M34986.1 | - | 7 - 8 | 912 | 60 | |
M60459.1 | - | 7 - 8 | 1017 | 60 |