Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLINT1
Entrez Gene ID
9685
Gene Name
clathrin interactor 1
Gene Aliases
CLINT, ENTH, EPN4, EPNR
Chromosome Location
Chr.5: 157785743 - 157859175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195555.1 NP_001182484.1 7 - 8 1147 71
NM_001195556.1 NP_001182485.1 7 - 8 1124 71
NM_014666.3 NP_055481.1 7 - 8 1147 71
XM_011534701.2 XP_011533003.1 7 - 8 1132 71
XM_017010087.1 XP_016865576.1 7 - 8 1132 71
XM_017010088.1 XP_016865577.1 7 - 8 1132 71
GenBank mRNA AF434813.1 - 7 - 8 942 71
AK092765.1 - 7 - 8 1094 71
AK300257.1 - 7 - 8 1115 71
BC004467.2 - 7 - 8 1008 71
D79993.1 - 7 - 8 1043 71
KU178486.1 - 7 - 8 942 71
KU178487.1 - 7 - 8 942 71

View Full Product Details