The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199453.3 | NP_955525.1 | 5 - 6 | 1126 | 73 |
GenBank mRNA | AJ011371.1 | - | 5 - 6 | 1081 | 73 |
AJ131725.1 | - | 5 - 6 | 1085 | 73 | |
AJ278981.1 | - | 5 - 6 | 1085 | 73 | |
AJ633645.1 | - | 5 - 6 | 1079 | 73 |