The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 10 - 11 | 2847 | 64 |
NM_181050.2 | NP_851393.1 | 9 - 10 | 2739 | 64 | |
NR_134879.1 | 8 - 9 | 2642 | 64 | ||
XM_011522682.2 | XP_011520984.1 | 10 - 11 | 2873 | 64 | |
XM_011522683.2 | XP_011520985.1 | 9 - 10 | 2764 | 64 | |
XM_011522684.2 | XP_011520986.1 | 10 - 11 | 2723 | 64 | |
XM_011522686.1 | XP_011520988.1 | 9 - 10 | 1630 | 64 | |
XM_017023743.1 | XP_016879232.1 | 10 - 11 | 2858 | 64 | |
XM_017023744.1 | XP_016879233.1 | 9 - 10 | 2749 | 64 | |
XM_017023745.1 | XP_016879234.1 | 9 - 10 | 1898 | 64 | |
XM_017023746.1 | XP_016879235.1 | 9 - 10 | 2474 | 64 | |
GenBank mRNA | AB208876.1 | - | 7 - 8 | 5528 | 64 |
AF009674.1 | - | 10 - 11 | 2573 | 64 | |
BC017447.1 | - | 5 - 6 | 1049 | 64 | |
BC035872.1 | - | 9 - 10 | 1888 | 64 | |
BC044648.1 | - | 9 - 10 | 2511 | 64 | |
BX648449.1 | - | 1 - 2 | 975 | 64 | |
KF615847.1 | - | 10 - 11 | 1818 | 64 | |
KF615848.1 | - | 9 - 10 | 1710 | 64 | |
KF615849.1 | - | 6 - 7 | 1334 | 64 | |
KF615851.1 | - | 5 - 6 | 1666 | 64 | |
KF615852.1 | - | 8 - 9 | 2642 | 64 | |
KF615853.1 | - | 5 - 6 | 716 | 64 | |
KF615854.1 | - | 5 - 6 | 563 | 64 | |
KU178413.1 | - | 8 - 9 | 2350 | 64 | |
KU178414.1 | - | 9 - 10 | 2458 | 64 | |
KU178415.1 | - | 7 - 8 | 2209 | 64 |