The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 7 - 8 | 2345 | 121 |
NM_181050.2 | NP_851393.1 | 7 - 8 | 2345 | 121 | |
NR_134879.1 | 6 - 7 | 2248 | 121 | ||
XM_011522682.2 | XP_011520984.1 | 7 - 8 | 2371 | 121 | |
XM_011522683.2 | XP_011520985.1 | 7 - 8 | 2370 | 121 | |
XM_011522684.2 | XP_011520986.1 | 7 - 8 | 2221 | 121 | |
XM_011522686.1 | XP_011520988.1 | 6 - 7 | 1128 | 121 | |
XM_017023743.1 | XP_016879232.1 | 7 - 8 | 2356 | 121 | |
XM_017023744.1 | XP_016879233.1 | 7 - 8 | 2355 | 121 | |
XM_017023745.1 | XP_016879234.1 | 6 - 7 | 1396 | 121 | |
XM_017023746.1 | XP_016879235.1 | 6 - 7 | 1972 | 121 | |
GenBank mRNA | AB208876.1 | - | 6 - 7 | 1253 | 121 |
BC017447.1 | - | 2 - 3 | 547 | 121 | |
BC035872.1 | - | 6 - 7 | 1386 | 121 | |
BC044648.1 | - | 7 - 8 | 2117 | 121 | |
KF615847.1 | - | 7 - 8 | 1316 | 121 | |
KF615848.1 | - | 7 - 8 | 1316 | 121 | |
KF615849.1 | - | 4 - 5 | 940 | 121 | |
KF615852.1 | - | 6 - 7 | 2248 | 121 | |
KU178413.1 | - | 6 - 7 | 1956 | 121 | |
KU178414.1 | - | 6 - 7 | 1956 | 121 | |
KU178415.1 | - | 5 - 6 | 1815 | 121 |