The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000440.2 | NP_000431.2 | 3 - 4 | 836 | 88 |
GenBank mRNA | BC035909.1 | - | 3 - 4 | 823 | 88 |
BC144044.1 | - | 3 - 4 | 763 | 88 | |
M26061.1 | - | 3 - 4 | 836 | 88 |