Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLIS2
Entrez Gene ID
84662
Gene Name
GLIS family zinc finger 2
Gene Aliases
NKL, NPHP7
Chromosome Location
Chr.16: 4314761 - 4339597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318918.1 NP_001305847.1 5 - 6 1342 62
NM_032575.2 NP_115964.2 3 - 4 578 62
XM_005255641.4 XP_005255698.1 4 - 5 643 62
GenBank mRNA AF325914.1 - 3 - 4 578 62
AK126918.1 - 5 - 6 1342 62

View Full Product Details