Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L3
Entrez Gene ID
23136
Gene Name
erythrocyte membrane protein band 4.1 like 3
Gene Aliases
4.1B, DAL-1, DAL1
Chromosome Location
Chr.18: 5392381 - 5630666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281533.1 NP_001268462.1 11 - 12 1567 130
NM_001281534.1 NP_001268463.1 11 - 12 1748 130
NM_001281535.1 NP_001268464.1 13 - 14 1549 130
XM_011525623.2 XP_011523925.1 11 - 12 1748 130
XM_011525626.2 XP_011523928.1 10 - 11 1350 130
XM_011525628.2 XP_011523930.1 11 - 12 1748 130
XM_011525632.2 XP_011523934.1 10 - 11 1350 130
XM_011525634.2 XP_011523936.1 10 - 11 1350 130
XM_011525637.2 XP_011523939.1 10 - 11 1350 130
XM_017025618.1 XP_016881107.1 10 - 11 1350 130
XM_017025619.1 XP_016881108.1 10 - 11 1350 130
XM_017025620.1 XP_016881109.1 10 - 11 1350 130
XM_017025621.1 XP_016881110.1 10 - 11 1350 130
XM_017025623.1 XP_016881112.1 10 - 11 1350 130
XM_017025624.1 XP_016881113.1 10 - 11 1350 130
XM_017025625.1 XP_016881114.1 11 - 12 1516 130
XM_017025626.1 XP_016881115.1 10 - 11 1350 130
XM_017025627.1 XP_016881116.1 11 - 12 1748 130
XM_017025628.1 XP_016881117.1 11 - 12 1748 130
XM_017025631.1 XP_016881120.1 10 - 11 1350 130
XM_017025632.1 XP_016881121.1 11 - 12 1748 130
XM_017025633.1 XP_016881122.1 11 - 12 1516 130
XM_017025634.1 XP_016881123.1 10 - 11 1350 130
XM_017025635.1 XP_016881124.1 10 - 11 1350 130
XM_017025636.1 XP_016881125.1 11 - 12 1748 130
XM_017025637.1 XP_016881126.1 11 - 12 1748 130
XM_017025638.1 XP_016881127.1 10 - 11 1350 130
XM_017025640.1 XP_016881129.1 11 - 12 1748 130
XM_017025641.1 XP_016881130.1 10 - 11 1350 130
XM_017025643.1 XP_016881132.1 11 - 12 1748 130
XM_017025645.1 XP_016881134.1 11 - 12 1607 130
XM_017025646.1 XP_016881135.1 11 - 12 1748 130
XM_017025647.1 XP_016881136.1 11 - 12 1607 130
XM_017025648.1 XP_016881137.1 11 - 12 1607 130
XM_017025650.1 XP_016881139.1 11 - 12 1607 130
XM_017025651.1 XP_016881140.1 11 - 12 1607 130
XM_017025654.1 XP_016881143.1 11 - 12 1607 130
XM_017025656.1 XP_016881145.1 11 - 12 1607 130
XM_017025657.1 XP_016881146.1 11 - 12 1607 130
XM_017025659.1 XP_016881148.1 11 - 12 1607 130
GenBank mRNA AF069072.1 - 11 - 12 1405 130
AK128531.1 - 12 - 13 1416 130
AK292583.1 - 13 - 14 1549 130
AK297406.1 - 11 - 12 1567 130
AK314438.1 - 9 - 10 1075 130
BC006141.1 - 11 - 12 1719 130

View Full Product Details