Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
Chromosome Location
Chr.9: 37509920 - 37576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012166.2 NP_036298.2 4 - 5 1613 68
XM_005251439.4 XP_005251496.1 4 - 5 1702 68
XM_006716754.3 XP_006716817.1 2 - 3 168 68
XM_017014618.1 XP_016870107.1 4 - 5 1819 68
XM_017014619.1 XP_016870108.1 4 - 5 1930 68
GenBank mRNA AB209610.1 - 3 - 4 1201 68
BC125124.1 - 2 - 3 207 68
BC125125.1 - 4 - 5 1692 68
BC140785.1 - 3 - 4 1584 68
BC171785.1 - 3 - 4 1584 68

View Full Product Details