Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
Chromosome Location
Chr.9: 37509920 - 37576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012166.2 NP_036298.2 7 - 8 1982 76
XM_006716754.3 XP_006716817.1 5 - 6 537 76
XM_017014618.1 XP_016870107.1 7 - 8 2188 76
XM_017014619.1 XP_016870108.1 7 - 8 2299 76
GenBank mRNA AB209610.1 - 6 - 7 1570 76
BC125124.1 - 5 - 6 576 76
BC125125.1 - 7 - 8 2061 76
BC140785.1 - 6 - 7 1953 76
BC171785.1 - 6 - 7 1953 76

View Full Product Details