The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022478.3 | NP_071923.2 | 6 - 7 | 1233 | 75 |
NM_144985.3 | NP_659422.2 | 6 - 7 | 1233 | 75 | |
XM_011537089.1 | XP_011535391.1 | 6 - 7 | 1314 | 75 | |
GenBank mRNA | AK057922.1 | - | 6 - 7 | 1169 | 75 |
AK289576.1 | - | 6 - 7 | 1233 | 75 | |
AK307746.1 | - | 6 - 7 | 1214 | 75 | |
AY260900.1 | - | 5 - 6 | 1066 | 75 | |
AY260901.1 | - | 5 - 6 | 1066 | 75 | |
AY358199.1 | - | 6 - 7 | 1189 | 75 | |
BX248303.1 | - | 2 - 3 | 257 | 75 |