Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRZ1
Entrez Gene ID
5803
Gene Name
protein tyrosine phosphatase, receptor type Z1
Gene Aliases
HPTPZ, HPTPzeta, PTP-ZETA, PTP18, PTPRZ, PTPZ, R-PTP-zeta-2, RPTPB, RPTPbeta, phosphacan
Chromosome Location
Chr.7: 121873105 - 122062036 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206838.1 NP_001193767.1 29 - 30 4624 63
NM_001206839.1 NP_001193768.1 28 - 29 4603 63
NM_002851.2 NP_002842.2 29 - 30 7204 63
XM_005250519.1 XP_005250576.1 28 - 29 7183 63
XM_017012477.1 XP_016867966.1 30 - 31 7251 63
GenBank mRNA AK294062.1 - 18 - 19 2458 63
CX872418.1 - 2 - 3 200 63
M93426.1 - 29 - 30 6953 63

View Full Product Details