The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003917.4 | NP_003908.1 | 5 - 6 | 672 | 72 |
XM_005268172.3 | XP_005268229.1 | 5 - 6 | 623 | 72 | |
XM_005268173.3 | XP_005268230.1 | 5 - 6 | 623 | 72 | |
XM_005268177.3 | XP_005268234.1 | 4 - 5 | 650 | 72 | |
XM_005268178.3 | XP_005268235.1 | 4 - 5 | 541 | 72 | |
XM_005268179.3 | XP_005268236.1 | 4 - 5 | 646 | 72 | |
XM_017021744.1 | XP_016877233.1 | 5 - 6 | 624 | 72 | |
XM_017021745.1 | XP_016877234.1 | 5 - 6 | 624 | 72 | |
XM_017021747.1 | XP_016877236.1 | 4 - 5 | 1909 | 72 | |
GenBank mRNA | AB015318.1 | - | 5 - 6 | 614 | 72 |
AF068706.1 | - | 4 - 5 | 1332 | 72 | |
BI918958.1 | - | 5 - 6 | 677 | 72 |