Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
Chromosome Location
Chr.14: 23559563 - 23569056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282474.1 NP_001269403.1 5 - 6 894 84
NM_001282475.1 NP_001269404.1 5 - 6 744 84
NM_003917.4 NP_003908.1 7 - 8 848 84
XM_005268167.3 XP_005268224.1 6 - 7 971 84
XM_005268168.4 XP_005268225.1 6 - 7 1013 84
XM_005268169.3 XP_005268226.1 6 - 7 970 84
XM_005268170.3 XP_005268227.1 6 - 7 970 84
XM_005268172.3 XP_005268229.1 7 - 8 799 84
XM_005268173.3 XP_005268230.1 7 - 8 799 84
XM_005268174.3 XP_005268231.1 5 - 6 846 84
XM_005268175.3 XP_005268232.1 5 - 6 860 84
XM_005268177.3 XP_005268234.1 6 - 7 826 84
XM_005268178.3 XP_005268235.1 6 - 7 717 84
XM_005268179.3 XP_005268236.1 6 - 7 822 84
XM_005268180.4 XP_005268237.1 5 - 6 1064 84
XM_005268182.3 XP_005268239.1 6 - 7 693 84
XM_006720301.3 XP_006720364.1 6 - 7 1068 84
XM_011537283.2 XP_011535585.1 5 - 6 917 84
XM_017021740.1 XP_016877229.1 5 - 6 917 84
XM_017021741.1 XP_016877230.1 3 - 4 821 84
XM_017021742.1 XP_016877231.1 5 - 6 888 84
XM_017021743.1 XP_016877232.1 5 - 6 917 84
XM_017021744.1 XP_016877233.1 7 - 8 800 84
XM_017021745.1 XP_016877234.1 7 - 8 800 84
XM_017021746.1 XP_016877235.1 6 - 7 693 84
XM_017021747.1 XP_016877236.1 6 - 7 2085 84
XM_017021748.1 XP_016877237.1 5 - 6 586 84
GenBank mRNA AB015318.1 - 7 - 8 790 84
AF068706.1 - 6 - 7 1508 84
AK304682.1 - 5 - 6 642 84
AK309103.1 - 5 - 6 1085 84
BC051833.1 - 4 - 5 685 84
BX161425.1 - 5 - 6 827 84
BX247963.1 - 4 - 5 553 84

View Full Product Details