Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
Chromosome Location
Chr.14: 23559563 - 23569056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282474.1 NP_001269403.1 17 - 18 2211 71
NM_001282475.1 NP_001269404.1 17 - 18 1999 71
NM_003917.4 NP_003908.1 19 - 20 2103 71
XM_005268167.3 XP_005268224.1 18 - 19 2226 71
XM_005268168.4 XP_005268225.1 18 - 19 2268 71
XM_005268169.3 XP_005268226.1 18 - 19 2225 71
XM_005268170.3 XP_005268227.1 18 - 19 2225 71
XM_005268172.3 XP_005268229.1 19 - 20 2054 71
XM_005268173.3 XP_005268230.1 19 - 20 2054 71
XM_005268174.3 XP_005268231.1 17 - 18 2101 71
XM_005268175.3 XP_005268232.1 17 - 18 2115 71
XM_005268177.3 XP_005268234.1 18 - 19 2081 71
XM_005268178.3 XP_005268235.1 18 - 19 1972 71
XM_005268179.3 XP_005268236.1 18 - 19 2077 71
XM_005268180.4 XP_005268237.1 17 - 18 2319 71
XM_005268182.3 XP_005268239.1 18 - 19 1948 71
XM_006720301.3 XP_006720364.1 18 - 19 2323 71
XM_011537283.2 XP_011535585.1 16 - 17 2067 71
XM_011537286.2 XP_011535588.1 10 - 11 1347 71
XM_017021740.1 XP_016877229.1 17 - 18 2172 71
XM_017021741.1 XP_016877230.1 15 - 16 2076 71
XM_017021742.1 XP_016877231.1 17 - 18 2143 71
XM_017021746.1 XP_016877235.1 18 - 19 2010 71
XM_017021747.1 XP_016877236.1 18 - 19 3402 71
XM_017021748.1 XP_016877237.1 16 - 17 2030 71
GenBank mRNA AB015318.1 - 19 - 20 2045 71
AF068706.1 - 18 - 19 2766 71
AK096034.1 - 4 - 5 2411 71
AK226158.1 - 9 - 10 1033 71
AK304682.1 - 17 - 18 1834 71
BC051833.1 - 16 - 17 1940 71
BX161425.1 - 17 - 18 2144 71

View Full Product Details