Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
Chromosome Location
Chr.14: 23559563 - 23569056 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005268182.3 XP_005268239.1 5 - 6 595 63
XM_017021746.1 XP_016877235.1 5 - 6 595 63
XM_017021748.1 XP_016877237.1 4 - 5 488 63
GenBank mRNA AK304682.1 - 4 - 5 544 63
BG701521.1 - 5 - 6 676 63
BX247963.1 - 3 - 4 455 63

View Full Product Details