Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35B4
Entrez Gene ID
84912
Gene Name
solute carrier family 35 member B4
Gene Aliases
YEA, YEA4
Chromosome Location
Chr.7: 134289337 - 134320307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032826.4 NP_116215.1 4 - 5 661 86
XM_011516645.2 XP_011514947.1 4 - 5 429 86
GenBank mRNA AB052892.1 - 4 - 5 637 86
AJ315497.1 - 4 - 5 493 86
AJ315498.1 - 4 - 5 483 86
AJ971941.1 - 4 - 5 337 86
AK027603.1 - 4 - 5 501 86
AK027726.1 - 3 - 4 219 86
AK075368.1 - 4 - 5 466 86
BC008413.1 - 4 - 5 482 86

View Full Product Details