The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021201.4 | NP_067024.1 | 5 - 6 | 735 | 69 |
NM_206938.1 | NP_996821.1 | 4 - 5 | 600 | 69 | |
NM_206939.1 | NP_996822.1 | 5 - 6 | 687 | 69 | |
NM_206940.1 | NP_996823.1 | 4 - 5 | 552 | 69 | |
GenBank mRNA | AB026043.1 | - | 5 - 6 | 675 | 69 |
AF201951.1 | - | 5 - 6 | 646 | 69 | |
AF237916.1 | - | 5 - 6 | 659 | 69 | |
AF309653.1 | - | 5 - 6 | 688 | 69 | |
AF350501.1 | - | 5 - 6 | 646 | 69 | |
AK075106.1 | - | 5 - 6 | 739 | 69 | |
AK297027.1 | - | 4 - 5 | 536 | 69 | |
BC020673.1 | - | 5 - 6 | 659 | 69 | |
CR457187.1 | - | 4 - 5 | 542 | 69 |