The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018518.4 | NP_060988.3 | 18 - 19 | 2622 | 73 |
NM_182751.2 | NP_877428.1 | 18 - 19 | 2625 | 73 | |
XM_011519538.2 | XP_011517840.1 | 18 - 19 | 2595 | 73 | |
GenBank mRNA | AB042719.1 | - | 18 - 19 | 2589 | 73 |
AF119869.1 | - | 7 - 8 | 961 | 73 | |
AK292701.1 | - | 18 - 19 | 2600 | 73 | |
AL136840.1 | - | 17 - 18 | 2588 | 73 | |
BC004876.2 | - | 11 - 12 | 1540 | 73 | |
BC009108.1 | - | 13 - 14 | 1840 | 73 | |
BC101727.1 | - | 18 - 19 | 2594 | 73 | |
BC143490.1 | - | 18 - 19 | 2587 | 73 |