The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021727.4 | NP_068373.1 | 2 - 3 | 495 | 60 |
XM_011545023.2 | XP_011543325.1 | 2 - 3 | 495 | 60 | |
XM_017017723.1 | XP_016873212.1 | 2 - 3 | 484 | 60 | |
XM_017017724.1 | XP_016873213.1 | 2 - 3 | 484 | 60 | |
GenBank mRNA | AB209356.1 | - | 1 - 2 | 2501 | 60 |
AF084560.1 | - | 2 - 3 | 464 | 60 | |
AF134404.1 | - | 2 - 3 | 401 | 60 | |
AK091608.1 | - | 1 - 2 | 669 | 60 | |
AK295473.1 | - | 2 - 3 | 426 | 60 | |
AK310523.1 | - | 2 - 3 | 403 | 60 | |
BC004901.2 | - | 2 - 3 | 484 | 60 |