Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
Chromosome Location
Chr.11: 61873523 - 61892224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021727.4 NP_068373.1 3 - 4 683 61
XM_011545023.2 XP_011543325.1 3 - 4 683 61
XM_017017723.1 XP_016873212.1 3 - 4 672 61
XM_017017724.1 XP_016873213.1 3 - 4 672 61
GenBank mRNA AB209356.1 - 2 - 3 2689 61
AF084560.1 - 3 - 4 652 61
AF134404.1 - 3 - 4 589 61
AK091608.1 - 2 - 3 857 61
AK295473.1 - 3 - 4 614 61
AK310523.1 - 3 - 4 591 61
BC004901.2 - 3 - 4 672 61

View Full Product Details