The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001873.3 | NP_001864.1 | 4 - 5 | 1068 | 92 |
GenBank mRNA | AK090962.1 | - | 5 - 6 | 852 | 92 |
AK127165.1 | - | 5 - 6 | 785 | 92 | |
AK290996.1 | - | 4 - 5 | 927 | 92 | |
AK294175.1 | - | 5 - 6 | 885 | 92 | |
BC033866.1 | - | 4 - 5 | 806 | 92 | |
BC053612.1 | - | 4 - 5 | 920 | 92 | |
X51405.1 | - | 4 - 5 | 1078 | 92 |