The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184694.2 | NP_001171623.1 | 3 - 4 | 191 | 97 |
NM_020203.3 | NP_064588.1 | 3 - 4 | 163 | 97 | |
GenBank mRNA | AF325916.1 | - | 3 - 4 | 144 | 97 |
AJ276396.1 | - | 3 - 4 | 154 | 97 | |
AK075076.1 | - | 3 - 4 | 191 | 97 | |
BC128158.1 | - | 3 - 4 | 154 | 97 |