Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEST1
Entrez Gene ID
7439
Gene Name
bestrophin 1
Gene Aliases
ARB, BEST, BMD, RP50, TU15B, VMD2
Chromosome Location
Chr.11: 61946722 - 61964465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004183.3 NP_004174.1 1 - 2 551 67
NR_134580.1 1 - 2 551 67
XM_005274210.3 XP_005274267.1 1 - 2 661 67
XM_005274219.3 XP_005274276.1 1 - 2 661 67
XM_005274221.3 XP_005274278.1 1 - 2 661 67
XM_017018229.1 XP_016873718.1 1 - 2 661 67
GenBank mRNA AF057169.1 - 1 - 2 75 67
AF057170.1 - 1 - 2 75 67
AF073501.1 - 1 - 2 59 67
AK289681.1 - 1 - 2 79 67
AK294577.1 - 1 - 2 79 67
AK295580.1 - 1 - 2 84 67

View Full Product Details