The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014207.3 | NP_055022.2 | 7 - 8 | 1330 | 116 |
GenBank mRNA | AK292698.1 | - | 7 - 8 | 1299 | 116 |
BC027901.1 | - | 7 - 8 | 1293 | 116 | |
X04391.1 | - | 7 - 8 | 1299 | 116 |