The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152716.2 | NP_689929.2 | 9 - 10 | 1268 | 63 |
XM_005273833.4 | XP_005273890.1 | 9 - 10 | 2390 | 63 | |
XM_005273834.1 | XP_005273891.1 | 8 - 9 | 1180 | 63 | |
XM_011544811.2 | XP_011543113.1 | 5 - 6 | 603 | 63 | |
XM_017017339.1 | XP_016872828.1 | 10 - 11 | 1344 | 63 | |
GenBank mRNA | AK094193.1 | - | 6 - 7 | 779 | 63 |
AK127943.1 | - | 8 - 9 | 1138 | 63 | |
AK293213.1 | - | 4 - 5 | 461 | 63 | |
AL831992.2 | - | 9 - 10 | 1209 | 63 | |
BC065264.1 | - | 6 - 7 | 762 | 63 | |
BC109038.1 | - | 6 - 7 | 723 | 63 | |
BC109039.1 | - | 6 - 7 | 723 | 63 | |
BC111047.1 | - | 7 - 8 | 930 | 63 |