The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 15 - 16 | 2139 | 99 |
XM_005270185.4 | XP_005270242.2 | 15 - 16 | 2179 | 99 | |
XM_011540213.1 | XP_011538515.1 | 15 - 16 | 2170 | 99 | |
XM_011540215.2 | XP_011538517.1 | 7 - 8 | 2364 | 99 | |
XM_017016696.1 | XP_016872185.1 | 15 - 16 | 2116 | 99 | |
XM_017016697.1 | XP_016872186.1 | 15 - 16 | 2132 | 99 | |
XM_017016698.1 | XP_016872187.1 | 15 - 16 | 1843 | 99 | |
XM_017016699.1 | XP_016872188.1 | 15 - 16 | 2123 | 99 | |
XM_017016700.1 | XP_016872189.1 | 15 - 16 | 1729 | 99 | |
GenBank mRNA | AF264912.1 | - | 15 - 16 | 1842 | 99 |
AF305081.1 | - | 15 - 16 | 2141 | 99 | |
AF309033.1 | - | 15 - 16 | 2093 | 99 | |
AF329696.1 | - | 15 - 16 | 1981 | 99 | |
AF342982.1 | - | 15 - 16 | 1842 | 99 | |
AF438201.1 | - | 15 - 16 | 2221 | 99 | |
AK314612.1 | - | 15 - 16 | 2107 | 99 | |
AL833676.1 | - | 1 - 2 | 177 | 99 |