The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 16 - 17 | 2354 | 86 |
XM_005270185.4 | XP_005270242.2 | 16 - 17 | 2394 | 86 | |
XM_011540213.1 | XP_011538515.1 | 16 - 17 | 2385 | 86 | |
XM_011540215.2 | XP_011538517.1 | 8 - 9 | 2579 | 86 | |
XM_017016696.1 | XP_016872185.1 | 16 - 17 | 2331 | 86 | |
XM_017016697.1 | XP_016872186.1 | 16 - 17 | 2347 | 86 | |
XM_017016698.1 | XP_016872187.1 | 16 - 17 | 2058 | 86 | |
XM_017016699.1 | XP_016872188.1 | 16 - 17 | 2338 | 86 | |
XM_017016700.1 | XP_016872189.1 | 16 - 17 | 1944 | 86 | |
GenBank mRNA | AF264912.1 | - | 16 - 17 | 2057 | 86 |
AF305081.1 | - | 16 - 17 | 2356 | 86 | |
AF309033.1 | - | 16 - 17 | 2308 | 86 | |
AF329696.1 | - | 16 - 17 | 2196 | 86 | |
AF342982.1 | - | 16 - 17 | 2057 | 86 | |
AF438201.1 | - | 16 - 17 | 2436 | 86 | |
AK314612.1 | - | 16 - 17 | 2322 | 86 | |
AL833676.1 | - | 2 - 3 | 392 | 86 |