The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 25 - 26 | 3584 | 75 |
XM_005270185.4 | XP_005270242.2 | 25 - 26 | 3624 | 75 | |
XM_011540213.1 | XP_011538515.1 | 25 - 26 | 3615 | 75 | |
XM_011540215.2 | XP_011538517.1 | 17 - 18 | 3809 | 75 | |
XM_017016696.1 | XP_016872185.1 | 25 - 26 | 3561 | 75 | |
XM_017016697.1 | XP_016872186.1 | 25 - 26 | 3577 | 75 | |
XM_017016698.1 | XP_016872187.1 | 25 - 26 | 3288 | 75 | |
XM_017016699.1 | XP_016872188.1 | 25 - 26 | 3568 | 75 | |
XM_017016700.1 | XP_016872189.1 | 25 - 26 | 3174 | 75 | |
GenBank mRNA | AF264912.1 | - | 25 - 26 | 3287 | 75 |
AF305081.1 | - | 25 - 26 | 3586 | 75 | |
AF309033.1 | - | 25 - 26 | 3538 | 75 | |
AF329696.1 | - | 25 - 26 | 3426 | 75 | |
AF342982.1 | - | 25 - 26 | 3287 | 75 | |
AF438201.1 | - | 25 - 26 | 3666 | 75 | |
AK023746.1 | - | 7 - 8 | 778 | 75 | |
AK314612.1 | - | 25 - 26 | 3552 | 75 | |
AL833676.1 | - | 11 - 12 | 1622 | 75 |