The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 5 - 6 | 928 | 66 |
XM_005270185.4 | XP_005270242.2 | 5 - 6 | 959 | 66 | |
XM_011540213.1 | XP_011538515.1 | 5 - 6 | 959 | 66 | |
XM_017016696.1 | XP_016872185.1 | 5 - 6 | 896 | 66 | |
XM_017016697.1 | XP_016872186.1 | 5 - 6 | 912 | 66 | |
XM_017016698.1 | XP_016872187.1 | 5 - 6 | 623 | 66 | |
XM_017016699.1 | XP_016872188.1 | 5 - 6 | 912 | 66 | |
XM_017016700.1 | XP_016872189.1 | 5 - 6 | 509 | 66 | |
XM_017016701.1 | XP_016872190.1 | 5 - 6 | 959 | 66 | |
GenBank mRNA | AF264912.1 | - | 5 - 6 | 631 | 66 |
AF305081.1 | - | 5 - 6 | 933 | 66 | |
AF309033.1 | - | 5 - 6 | 882 | 66 | |
AF329696.1 | - | 5 - 6 | 770 | 66 | |
AF342982.1 | - | 5 - 6 | 631 | 66 | |
AF438201.1 | - | 5 - 6 | 1010 | 66 | |
AK314612.1 | - | 5 - 6 | 896 | 66 |