The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303402.1 | NP_001290331.1 | 10 - 11 | 1129 | 126 |
NM_001303403.1 | NP_001290332.1 | 11 - 12 | 1181 | 126 | |
NM_020839.3 | NP_065890.1 | 11 - 12 | 1208 | 126 | |
XM_005265346.3 | XP_005265403.1 | 11 - 12 | 1208 | 126 | |
XM_011533971.1 | XP_011532273.1 | 11 - 12 | 1320 | 126 | |
XM_017006960.1 | XP_016862449.1 | 11 - 12 | 1320 | 126 | |
GenBank mRNA | AB040882.2 | - | 11 - 12 | 1179 | 126 |
AF468833.1 | - | 11 - 12 | 1168 | 126 | |
AK025513.1 | - | 11 - 12 | 1178 | 126 | |
AK074278.1 | - | 2 - 3 | 209 | 126 | |
AK297349.1 | - | 10 - 11 | 1099 | 126 | |
AK302307.1 | - | 11 - 12 | 1151 | 126 | |
AL832926.1 | - | 11 - 12 | 1162 | 126 | |
BC026353.1 | - | 11 - 12 | 1178 | 126 | |
BC037168.2 | - | 10 - 11 | 1044 | 126 |