Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR48
Entrez Gene ID
57599
Gene Name
WD repeat domain 48
Gene Aliases
P80, SPG60, UAF1
Chromosome Location
Chr.3: 39051986 - 39096671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303402.1 NP_001290331.1 13 - 14 1435 86
NM_001303403.1 NP_001290332.1 14 - 15 1487 86
NM_020839.3 NP_065890.1 14 - 15 1514 86
XM_005265346.3 XP_005265403.1 14 - 15 1514 86
XM_011533971.1 XP_011532273.1 14 - 15 1626 86
XM_017006960.1 XP_016862449.1 14 - 15 1626 86
GenBank mRNA AB040882.2 - 14 - 15 1485 86
AF468833.1 - 14 - 15 1474 86
AK025513.1 - 14 - 15 1484 86
AK074278.1 - 5 - 6 515 86
AK297349.1 - 13 - 14 1405 86
AK298810.1 - 6 - 7 659 86
AK302307.1 - 14 - 15 1457 86
AL832926.1 - 14 - 15 1468 86
BC026353.1 - 14 - 15 1484 86
BC037168.2 - 13 - 14 1350 86

View Full Product Details