Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR48
Entrez Gene ID
57599
Gene Name
WD repeat domain 48
Gene Aliases
P80, SPG60, UAF1
Chromosome Location
Chr.3: 39051986 - 39096671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303402.1 NP_001290331.1 15 - 16 1629 88
NM_001303403.1 NP_001290332.1 16 - 17 1681 88
NM_020839.3 NP_065890.1 16 - 17 1708 88
XM_005265346.3 XP_005265403.1 16 - 17 1708 88
XM_011533971.1 XP_011532273.1 16 - 17 1820 88
XM_017006960.1 XP_016862449.1 16 - 17 1820 88
GenBank mRNA AB040882.2 - 16 - 17 1679 88
AF468833.1 - 16 - 17 1668 88
AK025513.1 - 16 - 17 1678 88
AK074278.1 - 7 - 8 709 88
AK297349.1 - 15 - 16 1599 88
AK298810.1 - 8 - 9 853 88
AK302307.1 - 16 - 17 1651 88
AL162064.1 - 3 - 4 210 88
AL832926.1 - 16 - 17 1662 88
BC026353.1 - 16 - 17 1678 88
BC037168.2 - 15 - 16 1544 88

View Full Product Details