The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100819.2 | NP_001094289.1 | 2 - 3 | 206 | 70 |
NM_001204094.1 | NP_001191023.1 | 3 - 4 | 223 | 70 | |
NM_015387.4 | NP_056202.2 | 3 - 4 | 269 | 70 | |
NM_199482.3 | NP_955776.1 | 3 - 4 | 279 | 70 | |
GenBank mRNA | AB015441.1 | - | 3 - 4 | 228 | 70 |
AF093825.1 | - | 3 - 4 | 244 | 70 | |
AF151853.1 | - | 3 - 4 | 244 | 70 | |
AF250319.1 | - | 3 - 4 | 254 | 70 | |
AJ580638.1 | - | 3 - 4 | 225 | 70 | |
AK027043.1 | - | 3 - 4 | 266 | 70 | |
AK289377.1 | - | 3 - 4 | 259 | 70 | |
AK292938.1 | - | 3 - 4 | 265 | 70 | |
AK297514.1 | - | 2 - 3 | 165 | 70 | |
AL080070.1 | - | 3 - 4 | 238 | 70 | |
BC005237.1 | - | 3 - 4 | 269 | 70 | |
CR457371.1 | - | 3 - 4 | 225 | 70 | |
DC298610.1 | - | 3 - 4 | 279 | 70 | |
DC350915.1 | - | 2 - 3 | 202 | 70 | |
DC375649.1 | - | 3 - 4 | 225 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202485.1 | NP_001189414.1 | 4 - 5 | 773 | 70 |
GenBank mRNA | BJ992310.1 | - | 4 - 5 | 391 | 70 |
CR990871.1 | - | 4 - 5 | 357 | 70 |